L-Glutamic acid, [14C(U)] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC156-50UCI MC156-250UCI MC156-1MCI
101970-704EA 5387.69 USD
101970-704 101970-706 101970-708
L-Glutamic acid, [14C(U)] ≥97%
L-Glutamic acid, [14C(U)]
CAS Number: 2088140-92-3
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR