2'-Deoxyuridine-5'-monophosphate, diammonium salt, [2-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC153-10UCI MC153-50UCI
101970-688EA 1693.46 USD
101970-688 101970-692
2'-Deoxyuridine-5'-monophosphate, diammonium salt, [2-14C] ≥97% (by HPLC)
2'-Deoxyuridine-5'-monophosphate, diammonium salt, [2-14C]
CAS Number: 335021-67-5
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR