D-Glucose-6-phosphate, [14C(U)] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC149-50UCI MC149-250UCI
101970-674EA 5387.69 USD
101970-674 101970-676
D-Glucose-6-phosphate, [14C(U)] ≥97% (by HPLC)
D-Glucose-6-phosphate, [14C(U)]
CAS Number: 2089110-62-1
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR