D-Fructose-6-phosphate, disodium salt, [14C(U)] ≥95%

Supplier: MORAVEK BIOCHEMICALS MS

MC1462-50UCI MC1462-250UCI
101970-644EA 5387.69 USD
101970-644 101970-646
D-Fructose-6-phosphate, disodium salt, [14C(U)] ≥95%
D-Fructose-6-phosphate, disodium salt, [14C(U)]
CAS Number: 2088859-01-0
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR