Guanidine hydrochloride, [14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC145S-50UCI MC145S-250UCI
101970-632EA 5387.69 USD
101970-632 101970-634
Guanidine hydrochloride, [14C] ≥98% (by HPLC), MORpure™
Guanidine hydrochloride, [14C]
CAS Number: 73549-39-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR