D-Fructose, [14C(U)] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC1459-50UCI MC1459-250UCI MC1459-1MCI
101970-626EA 5387.69 USD
101970-626 101970-628 101970-630
D-Fructose, [14C(U)] ≥97% (by HPLC)
D-Fructose, [14C(U)]
CAS Number: 685-78-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR