Decanoic acid, [1-14C] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC1455-50UCI MC1455-250UCI
101970-612EA 5387.69 USD
101970-612 101970-614
Decanoic acid, [1-14C] ≥97%
Decanoic acid, [1-14C]
CAS Number: 94033-36-0
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR