Guanidine hydrochloride, [14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC145-50UCI MC145-250UCI
101970-604EA 5387.69 USD
101970-604 101970-606
Guanidine hydrochloride, [14C] ≥98%, MORpure™
Guanidine hydrochloride, [14C]
CAS Number: 73549-39-0
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR