L-Norleucine, [1-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1424-50UCI MC1424-250UCI
101970-538EA 5387.69 USD
101970-538 101970-540
L-Norleucine, [1-14C] ≥98%, MORpure™
L-Norleucine, [1-14C]
CAS Number: 288852-41-5
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR