2-Chloro-2'-deoxyadenosine, [8-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1418-10UCI MC1418-50UCI
101970-520EA 1693.46 USD
101970-520 101970-522
2-Chloro-2'-deoxyadenosine, [8-14C] ≥98% (by HPLC), MORpure™
2-Chloro-2'-deoxyadenosine, [8-14C]
CAS Number: 333382-22-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR