L-Aspartic acid, [14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC139H-50UCI MC139H-250UCI
101970-474EA 5387.69 USD
101970-474 101970-476
L-Aspartic acid, [14C(U)] ≥98%, MORpure™
L-Aspartic acid, [14C(U)]
CAS Number: 4076-01-1
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR