α-Methyl[14C]-L-tryptophan ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1396-10UCI MC1396-50UCI
101970-466EA 1693.46 USD
101970-466 101970-468
α-Methyl[14C]-L-tryptophan ≥98%, MORpure™
α-Methyl[14C]-L-tryptophan
CAS Number: 126313-95-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR