Ganciclovir, [8-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1392-10UCI MC1392-25UCI
101970-450EA 1693.46 USD
101970-450 101970-452
Ganciclovir, [8-14C] ≥98%, MORpure™
Ganciclovir, [8-14C]
CAS Number: 220199-26-8
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR