L-Aspartic acid, [14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC139-50UCI MC139-250UCI MC139-1MCI
101970-440EA 5191.54 USD
101970-440 101970-442 101970-444
L-Aspartic acid, [14C(U)] ≥98%, MORpure™
L-Aspartic acid, [14C(U)]
CAS Number: 4076-01-1
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR