Guanidinoacetic acid, [1-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1387-10UCI MC1387-50UCI
101970-430EA 1693.46 USD
101970-430 101970-432
Guanidinoacetic acid, [1-14C] ≥98%, MORpure™
Guanidinoacetic acid, [1-14C]
CAS Number: 333382-13-1
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR