Acyclovir, [8-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1386-10UCI MC1386-25UCI
101970-422EA 1693.46 USD
101970-422 101970-424
Acyclovir, [8-14C] ≥98%, MORpure™
Acyclovir, [8-14C]
CAS Number: 333382-12-0
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR