L-Arginine, [14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC137-50UCI MC137-250UCI
101970-400EA 5387.69 USD
101970-400 101970-402
L-Arginine, [14C(U)] ≥98%, MORpure™
L-Arginine, [14C(U)]
CAS Number: 2065186-04-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR