Potassium cyanate, [14C] ≥96% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC1339-250UCI MC1339-1MCI
101970-372EA 3615.77 USD
101970-372 101970-374
Potassium cyanate, [14C] ≥96% (by HPLC)
Potassium cyanate, [14C]
CAS Number: 67877-97-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR