Nicotinic acid, [carboxyl-14C] ≥97%

Supplier: MORAVEK BIOCHEMICALS MS

MC1324-250UCI MC1324-1MCI
101970-342EA 3386.92 USD
101970-342 101970-344
Nicotinic acid, [carboxyl-14C] ≥97%
Nicotinic acid, [carboxyl-14C]
CAS Number: 2906-42-5
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR