L-Alanine, [14C(U)] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC132-50UCI MC132-250UCI MC132-1MCI
101970-336EA 5387.69 USD
101970-336 101970-338 101970-340
L-Alanine, [14C(U)] ≥98% (by HPLC), MORpure™
L-Alanine, [14C(U)]
CAS Number: 10098-48-3
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR