Adenine, [14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC129-50UCI MC129-250UCI
101970-314EA 5387.69 USD
101970-314 101970-316
Adenine, [14C(U)] ≥98%, MORpure™
Adenine, [14C(U)]
CAS Number: 2088576-51-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR