Cytidine, [2-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC128-50UCI MC128-100UCI
101970-310EA 5387.69 USD
101970-310 101970-312
Cytidine, [2-14C] ≥98% (by HPLC), MORpure™
Cytidine, [2-14C]
CAS Number: 74203-88-6
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR