Benzoic acid, [ring-14C(U)] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1252S-1MCI MC1252S-5MCI
101970-266EA 6368.46 USD
101970-266 101970-268
Benzoic acid, [ring-14C(U)] ≥98%, MORpure™
Benzoic acid, [ring-14C(U)]
CAS Number: 74920-45-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR