Benzoic acid, [ring-14C(U)] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1252-100UCI MC1252-250UCI MC1252-1MCI MC1252-5MCI
101970-258EA 4472.31 USD
101970-258 101970-260 101970-262 101970-264
Benzoic acid, [ring-14C(U)] ≥98% (by HPLC), MORpure™
Benzoic acid, [ring-14C(U)]
CAS Number: 74920-45-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR