2'-Deoxyuridine, [2-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC119-10UCI MC119-50UCI MC119-250UCI
101970-190EA 1693.46 USD
101970-190 101970-192 101970-194
2'-Deoxyuridine, [2-14C] ≥98% (by HPLC), MORpure™
2'-Deoxyuridine, [2-14C]
CAS Number: 17236-65-6
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR