5-Fluoro-2'-deoxyuridine 5'-monophosphate, diammonium salt, [2-14C] ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

MC113-10UCI MC113-50UCI
101970-128EA 1693.46 USD
101970-128 101970-130
5-Fluoro-2'-deoxyuridine 5'-monophosphate, diammonium salt, [2-14C] ≥97% (by HPLC)
5-Fluoro-2'-deoxyuridine 5'-monophosphate, diammonium salt, [2-14C]
CAS Number: 332345-06-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR