Myristic acid, [1-14C] ≥98% (by HPLC), MORpure™

MC1118-50UCI MC1118-250UCI
101970-112EA 5387.69 USD
101970-112 101970-114
Myristic acid, [1-14C] ≥98% (by HPLC), MORpure™
Myristic acid, [1-14C]
CAS Number: 17029-30-0
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR