Uridine, [2-14C] ≥98% (by HPLC), MORpure™

MC105-10UCI MC105-50UCI MC105-250UCI
101970-080EA 1693.46 USD
101970-080 101970-082 101970-084
Uridine, [2-14C] ≥98% (by HPLC), MORpure™
Uridine, [2-14C]
CAS Number: 54-23-9
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR