5-Fluoro-2'-deoxyuridine, [2-14C] ≥98% (by HPLC), MORpure™

MC103-10UCI MC103-50UCI
101970-068EA 1693.46 USD
101970-068 101970-070
5-Fluoro-2'-deoxyuridine, [2-14C] ≥98% (by HPLC), MORpure™
5-Fluoro-2'-deoxyuridine, [2-14C]
CAS Number: 26291-67-8
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR