Penciclovir

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 2-Amino-1,9-dihydro-9-[4-hydroxy-3-(hydroxymethyl)butyl]-6H-purin-6-one, 2-Amino-9-[4-hydroxy-3-(hydroxymethyl)butyl]-1H-purin-6-one, 9-[4-Hydroxy-3-(hydroxymethyl)butyl]guanine, BRL 39123, 2-Amino-9-(4-hydroxy-3-(hydroxymethyl)butyl)-1H-purin-6(9H)-one

M972-1MG M972-5MG
101970-038EA 294.23 USD
101970-038 101970-040
Penciclovir
Penciclovir
Formula: C₁₀H₁₅N₅O₃
MW: 253.26 g/mol
MDL Number: MFCD15142866
CAS Number: 39809-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR