((2-(6-Amino-9H-purin-9-yl)ethoxy)-methyl)phosphonic acid

Supplier: MORAVEK BIOCHEMICALS MS

M703-1MG M703-5MG
101969-966EA 294.23 USD
101969-966 101969-968
((2-(6-Amino-9H-purin-9-yl)ethoxy)-methyl)phosphonic acid
((2-(6-Amino-9H-purin-9-yl)ethoxy)methyl)phosphonic acid
Formula: C₈H₁₂N₅O₄P
MDL Number: MFCD00866943
CAS Number: 106941-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR