β-L-Thymidine ≥97% (by HPLC)

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: Telbivudine, 1-((2S,4R,5S)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

M614-1MG M614-5MG
101969-948EA 294.23 USD
101969-948 101969-950
β-L-Thymidine ≥97% (by HPLC)
β-L-Thymidine
Formula: C₁₀H₁₄N₂O₅
MW: 242.2286 g/mol
MDL Number: MFCD02683612
CAS Number: 3424-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR