β-L-Guanosine ≥98% (by HPLC) MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: L-Guanosine, 2-Amino-9-((2S,3S,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

M323-1MG M323-5MG
101969-900EA 294.23 USD
101969-900 101969-902
β-L-Guanosine ≥98% (by HPLC) MORpure™
β-L-Guanosine
Formula: C₁₀H₁₃N₅O₅
MW: 283.2407 g/mol
MDL Number: MFCD00064103
CAS Number: 26578-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR