L-Phenylalanine
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
M1888-1MG
M1888-5MG
101969-814EA
294.23
USD
101969-814
101969-816
L-Phenylalanine
L-Phenylalanine
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol |
MDL Number:
MFCD00064227 CAS Number: 63-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...