Zoledronic acid ≥98% (by HPLC), MORpure™
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
(1-Hydroxy-2-(1H-imidazol-1-yl)ethane-1,1-diyl)diphosphonic acid, ZOL-446, CGP 4244, CGP 42446A, Zoledronate
M1875-1MG
M1875-5MG
101969-784EA
294.23
USD
101969-784
101969-786
Zoledronic acid ≥98% (by HPLC), MORpure™
Zoledronic acid
Formula:
C₅H₁₀N₂O₇P₂ MW: 272.09 g/mol |
MDL Number:
MFCD00867791 CAS Number: 118072-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...