amifostine
Supplier: MORAVEK BIOCHEMICALS MS
M1865-1MG
M1865-5MG
101969-756EA
294.23
USD
101969-756
101969-758
amifostine
Amifostine
Formula:
C₅H₁₅N₂O₃PS MW: 214.22 g/mol |
CAS Number:
20537-88-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...