Zanamivir hydrate ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: (2R,3R,4S)-3-acetamido-4-(diaminomethylideneamino)-2-[(1R,2R)-1,2,3-trihydroxypropyl]-3,4-dihydro-2H-pyran-6-carboxylic acid hydrate, 4-Guanidino-2,4-dideoxy-2,3-dehydro-N-acetylneuraminic acid hydrate, 4-Guanidino-Neu5Ac2en hydrate

M1826-1MG M1826-5MG
101969-736EA 294.23 USD
101969-736 101969-738
Zanamivir hydrate ≥98% (by HPLC), MORpure™
Zanamivir hydrate
Formula: C₁₂H₂₀N₄O₇·xH₂O
MW: 332.31 g/mol (anhy)
CAS Number: 551942-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR