Nelfinavir mesylate, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: (3S,4aS,8aS)-N-(tert-Butyl)-2-((2R,3R)-2-hydroxy-3-(3-hydroxy-2-methylbenzamido)-4-(phenylthio)butyl)decahydroisoquinoline-3-carboxamide methanesulphonate, AG 1341

M1824-1MG M1824-5MG
101969-732EA 294.23 USD
101969-732 101969-734
Nelfinavir mesylate, MORpure™
Nelfinavir mesylate
Formula: C₃₂H₄₅N₃O₄S·CH₄O₃S
MW: 663.9 g/mol
MDL Number: MFCD00931436
CAS Number: 159989-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR