2'-O-Methyluridine
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 2′-O-Methyluridine
M1815-1MG
M1815-5MG
101969-696EA
294.23
USD
101969-696
101969-698
2'-O-Methyluridine
2'-O-Methyluridine
Formula:
C₁₀H₁₄N₂O₆ MW: 258.23 g/mol |
MDL Number:
MFCD00056054 CAS Number: 2140-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...