N3-Methyluridine ≥98% (by HPLC) MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 3-Methyluridine, 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3-methylpyrimidine-2,4(1H,3H)-dione

M1804-1MG M1804-5MG
101969-656EA 294.23 USD
101969-656 101969-658
N3-Methyluridine ≥98% (by HPLC) MORpure™
N3-Methyluridine
Formula: C₁₀H₁₄N₂O₆
MW: 258.2280 g/mol
MDL Number: MFCD07778657
CAS Number: 2140-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR