5-Iodouridine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione

M1795-1MG M1795-5MG
101969-616EA 294.23 USD
101969-616 101969-618
5-Iodouridine
5-Iodouridine
Formula: C₉H₁₁IN₂O₆
MW: 370.10 g/mol
MDL Number: MFCD00006532
CAS Number: 1024-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR