5-Bromocytidine ≥98% (by HPLC) MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 4-Amino-5-bromo-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

M1789-1MG M1789-5MG
101969-596EA 294.23 USD
101969-596 101969-598
5-Bromocytidine ≥98% (by HPLC) MORpure™
5-Bromocytidine
Formula: C₉H₁₂BrN₃O₅
MW: 322.1127 g/mol
MDL Number: MFCD00057461
CAS Number: 3066-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR