2'-Deoxy-2'-fluorouridine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 2'-Fluoro-2'-deoxyuridine, 1-((2R,3R,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

M1786-1MG M1786-5MG
101969-584EA 294.23 USD
101969-584 101969-586
2'-Deoxy-2'-fluorouridine
2'-Deoxy-2'-fluorouridine
Formula: C₉H₁₁FN₂O₅
MW: 246.19 g/mol
MDL Number: MFCD01317293
CAS Number: 784-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR