7-Deaza-2'-deoxyguanosine ≥98% (by HPLC) MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 2-Amino-7-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-pyrrolo[2,3-d]pyrimidin-4(7H)-one

M1776-1MG M1776-5MG
101969-548EA 294.23 USD
101969-548 101969-550
7-Deaza-2'-deoxyguanosine ≥98% (by HPLC) MORpure™
7-Deaza-2'-deoxyguanosine
Formula: C₁₁H₁₄N₄O₄
MW: 266.2533 g/mol
MDL Number: MFCD00672278
CAS Number: 86392-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR