7-Deaza-2'-deoxyadenosine ≥98% (by HPLC) MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: (2R,3S,5R)-5-(4-Amino-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol

M1775-1MG M1775-5MG
101969-544EA 294.23 USD
101969-544 101969-546
7-Deaza-2'-deoxyadenosine ≥98% (by HPLC) MORpure™
7-Deaza-2'-deoxyadenosine
Formula: C₁₁H₁₄N₄O₃
MW: 250.2539 g/mol
MDL Number: MFCD00672266
CAS Number: 60129-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR