2'-Deoxy-5-methylcytidine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one, 5-mdC, 5-Methyl-2'-deoxycytidine, 1-(2-Deoxy-beta-D-ribofuranosyl)-5-methylcytosine

M1767-1MG M1767-5MG
101969-520EA 294.23 USD
101969-520 101969-522
2'-Deoxy-5-methylcytidine
2'-Deoxy-5-methylcytidine
Formula: C₁₀H₁₅N₃O₄
MW: 241.25 g/mol
MDL Number: MFCD00006549
CAS Number: 838-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR