8-Bromo-2'-deoxyguanosine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 2-Amino-8-bromo-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one

M1754-1MG M1754-5MG
101969-472EA 294.23 USD
101969-472 101969-474
8-Bromo-2'-deoxyguanosine
8-Bromo-2'-deoxyguanosine
Formula: C₁₀H₁₂BrN₅O₄
MW: 346.14 g/mol
MDL Number: MFCD01630971
CAS Number: 13389-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR