Ciprofloxacin

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid

M1737-1MG M1737-5MG
101969-416EA 294.23 USD
101969-416 101969-418
Ciprofloxacin
Ciprofloxacin
Formula: C₁₇H₁₈FN₃O₃
MW: 331.35 g/mol
MDL Number: MFCD00185755
CAS Number: 85721-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR