Lopinavir ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: Koletra, N-((2S,4S,5S)-5-(2-(2,6-Dimethylphenoxy)acetamido)-4-hydroxy-1,6-diphenylhexan-2-yl)-3-methyl-2-(2-oxotetrahydropyrimidin-1(2H)-yl)butanamide, ABT-378, Aluviran, A-157378.0, (S)-N-((2R,4S,5S)-5-(2-(2,6-Dimethylphenoxy)acetamido)-4-hydroxy-1,6-diphenylhexn-2-yl)-3-methyl-2-(2-oxotetrahydropyrimidin-1(2H)-yl)butanamide

M1735-1MG M1735-5MG
101969-404EA 294.23 USD
101969-404 101969-406
Lopinavir ≥98% (by HPLC), MORpure™
Lopinavir
Formula: C₃₇H₄₈N₄O₅
MW: 628.82 g/mol
MDL Number: MFCD22628840
CAS Number: 192725-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR