Famciclovir

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 2-[2-(2-Amino-9H-purin-9-yl)ethyl]-1,3-propanediol Diacetate, 2-(2-(2-Amino-9H-purin-9-yl)ethyl)propane-1,3-diyl diacetate

M1726-1MG M1726-5MG
101969-360EA 300.77 USD
101969-360 101969-362
Famciclovir
Famciclovir
Formula: C₁₄H₁₉N₅O₄
MW: 321.34 g/mol
MDL Number: MFCD00866964
CAS Number: 104227-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR